About This Item
Recommended Products
assay
98%
refractive index
n20/D 1.565 (lit.)
bp
273 °C (lit.)
density
1.388 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
Clc1ccc(CN=C=O)c(Cl)c1
InChI
1S/C8H5Cl2NO/c9-7-2-1-6(4-11-5-12)8(10)3-7/h1-3H,4H2
InChI key
FUIUATRKBRZCQD-UHFFFAOYSA-N
General description
application
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service