Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478199

Sigma-Aldrich

Dimethyl 5-isocyanatoisophthalate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNC6H3(CO2CH3)2
CAS Number:
Molecular Weight:
235.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

mp

106-109 °C (lit.)

SMILES string

COC(=O)c1cc(cc(c1)C(=O)OC)N=C=O

InChI

1S/C11H9NO5/c1-16-10(14)7-3-8(11(15)17-2)5-9(4-7)12-6-13/h3-5H,1-2H3

InChI key

JHCYWIFFIHXGAG-UHFFFAOYSA-N

General description

Dimethyl 5-isocyanatoisophthalate is an organic building block containing an isocyanate group.

Application

Dimethyl 5-isocyanatoisophthalate may be used as a starting material in the synthesis of dimethyl 5-(3-(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)ureido)isophthalate, a potential 14-3-3 protein-protein interaction (PPI) inhibitor.[1]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koushik Sarkar et al.
Chemistry, an Asian journal, 14(1), 194-204 (2018-10-26)
A structural rationale was adopted to design a series of metallogels from a newly synthesized urea-functionalized dicarboxylate ligand, namely, 5-[3-(pyridin-3-yl)ureido]isophthalic acid (PUIA), that produces metallogels upon reaction with various metal salts (CuII , ZnII , CoII , CdII , and
Discovery of 14-3-3 Protein-Protein Interaction Inhibitors that Sensitize Multidrug-Resistant Cancer Cells to Doxorubicin and the Akt Inhibitor GSK690693.
Mori M, et al.
ChemMedChem, 9(5), 973-983 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service