Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476498

Sigma-Aldrich

1,1,2-Tribromoethane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CHBr2
CAS Number:
Molecular Weight:
266.76
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.593 (lit.)

bp

187-190 °C/721 mmHg (lit.)

density

2.61 g/mL at 25 °C (lit.)

SMILES string

BrCC(Br)Br

InChI

1S/C2H3Br3/c3-1-2(4)5/h2H,1H2

InChI key

QUMDOMSJJIFTCA-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service