Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476137

Sigma-Aldrich

mono-Butyl maleate

90%

Synonym(s):

Butyl hydrogen maleate, Hydrogen butyl maleate, Monobutyl maleate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2(CH2)3CH3
CAS Number:
Molecular Weight:
172.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

90%

refractive index

n20/D 1.458 (lit.)

bp

201 °C (lit.)

mp

−1.5 °C (lit.)

density

1.099 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)\C=C/C(O)=O

InChI

1S/C8H12O4/c1-2-3-6-12-8(11)5-4-7(9)10/h4-5H,2-3,6H2,1H3,(H,9,10)/b5-4-

InChI key

UTOVMEACOLCUCK-PLNGDYQASA-N

Other Notes

remainder maleic acid and n-butanol

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanno C Erythropel et al.
Chemosphere, 134, 106-112 (2015-04-29)
The ubiquitous presence of the plasticizer di (2-ethylhexyl) phthalate (DEHP) in the environment is of concern due to negative biological effects associated with it and its metabolites. In particular, the metabolite mono (2-ethylhexyl) phthalate (MEHP) is a potential endocrine disruptor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service