Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476005

Sigma-Aldrich

2,3-Dichlorophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NCS
CAS Number:
Molecular Weight:
204.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.675 (lit.)

bp

256-258 °C (lit.)

density

1.434 g/mL at 25 °C (lit.)

SMILES string

Clc1cccc(N=C=S)c1Cl

InChI

1S/C7H3Cl2NS/c8-5-2-1-3-6(7(5)9)10-4-11/h1-3H

InChI key

RQZIODPVCCTBAQ-UHFFFAOYSA-N

General description

2,3-Dichlorophenyl isothiocyanate is an isothiocyanate derivative. Its freezing point has been reported to be 530.15K.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 135-135 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service