Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47472

Sigma-Aldrich

Fmoc-Phe(4-NO2)-OH

≥96.0% (HPLC)

Synonym(s):

Fmoc-4-nitro-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H20N2O6
CAS Number:
Molecular Weight:
432.43
Beilstein/REAXYS Number:
5178656
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥96.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1ccc(cc1)[N+]([O-])=O)NC(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C24H20N2O6/c27-23(28)22(13-15-9-11-16(12-10-15)26(30)31)25-24(29)32-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,25,29)(H,27,28)/t22-/m0/s1

InChI key

RZRRJPNDKJOLHI-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Bellavita et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 1751-1764 (2020-09-23)
The rapid development of antimicrobial resistance is pushing the search in the discovering of novel antimicrobial molecules to prevent and treat bacterial infections. Self-assembling antimicrobial peptides, as the lipidated peptides, are a novel and promising class of molecules capable of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service