Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

474703

Sigma-Aldrich

2,2′-Dimethylbiphenyl

97%

Synonym(s):

o,o′-Bitoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4C6H4CH3
CAS Number:
Molecular Weight:
182.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5745 (lit.)

density

0.989 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1-c2ccccc2C

InChI

1S/C14H14/c1-11-7-3-5-9-13(11)14-10-6-4-8-12(14)2/h3-10H,1-2H3

InChI key

ABMKWMASVFVTMD-UHFFFAOYSA-N

General description

2,2′-Dimethylbiphenyl is also referred to as o,o′-bitoluene. Preparation of optically active 2,2′-dimethylbiphenyl by using optically stable (+) or (-)-2,2′-dimethylbiphenyl-6,6′-diamine as a starting reagent has been reported.[1] Its electronic absorption spectrum has been reported.[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzuki H.
Electronic absorption spectra and geometry of organic molecules: An application of molecular orbital theory (Elsevier), 283-283 (2012)
Optically Active 2, 2'-Dimethylbiphenyl, the Simplest Atropisomeric Hydrocarbon.
Theilacker W and Bohm H.
Angewandte Chemie (International Edition in English), 6(3), 251-251 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service