Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

474541

Sigma-Aldrich

Nafion SAC-13

pore diameter >10 nm, pore volume >0.6 mL/g

Synonym(s):

Perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic acid-tetrafluoroethylene, Tetrafluoroethylene-perfluoro-3,6-dioxa-4-methyl-7-octenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203

composition

fluorosulfonic acid Nafion polymer on amorphous silica, 10-20% (porous nanocomposite)

pore size

>0.6 mL/g pore volume
>10 nm pore diameter

density

2.1 g/mL at 25 °C (lit.)

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

Inchi Key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Application

Strongly acidic catalyst resin.

Features and Benefits

Stable to 200°C. Catalyst for acylations, alkylations, esterifications and isomerizations. 10x4 times more surface area than Nafion pellets.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Ho Park et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4308-4314 (2015-06-24)
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service