Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47438

Sigma-Aldrich

Fmoc-Ala-OPfp

≥96.0% (HPLC)

Synonym(s):

Fmoc-L-alanine pentafluorophenyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16F5NO4
CAS Number:
Molecular Weight:
477.38
Beilstein/REAXYS Number:
3639513
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

assay

≥96.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

C[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(=O)Oc4c(F)c(F)c(F)c(F)c4F

InChI

1S/C24H16F5NO4/c1-11(23(31)34-22-20(28)18(26)17(25)19(27)21(22)29)30-24(32)33-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,30,32)/t11-/m0/s1

InChI key

CJXZXBGKOSXBFR-NSHDSACASA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service