Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470805

Sigma-Aldrich

2-Acetylphenylboronic acid

≥95%

Synonym(s):

2-Acetylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC6H4B(OH)2
CAS Number:
Molecular Weight:
163.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

mp

170 °C (dec.) (lit.)

SMILES string

CC(=O)c1ccccc1B(O)O

InChI

1S/C8H9BO3/c1-6(10)7-4-2-3-5-8(7)9(11)12/h2-5,11-12H,1H3

InChI key

ZKAOVABYLXQUTI-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki-Miyaura cross-coupling reactions
  • Rhodium-catalyzed annulation of ynamides
  • Transient masking of hydroxy groups
  • Cationic palladium complex-catalyzed diastereoselective tandem annulation
  • Hydrogen peroxide mediated formation of heteroaryl ethers
  • Copper-catalyzed transmetalation and homocoupling

Other Notes

Contains varying amounts of anhydride

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service