Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

470716

Sigma-Aldrich

2,2′-Oxydianiline

98%

Synonym(s):

2-Aminophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(C6H4NH2)2
CAS Number:
Molecular Weight:
200.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

61-64 °C (lit.)

SMILES string

Nc1ccccc1Oc2ccccc2N

InChI

1S/C12H12N2O/c13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)14/h1-8H,13-14H2

InChI key

GOJFAKBEASOYNM-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyimides derived from 2, 2'-bis (trifluoromethyl)-4, 4'-diaminobiphenyl. 3. Property control for polymer blends and copolymerization of fluorinated polyimides.
Matsuura T, et al.
Macromolecules, 26(3), 419-423 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service