Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

470392

Sigma-Aldrich

2-Fluoro-5-methylphenol

98%

Synonym(s):

6-Fluoro-m-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)OH
CAS Number:
Molecular Weight:
126.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

98%

refractive index

n20/D 1.511 (lit.)

bp

173 °C (lit.)

density

1.155 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(F)c(O)c1

InChI

1S/C7H7FO/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,1H3

InChI key

XEHPMVZYZDQLDN-UHFFFAOYSA-N

General description

2-Fluoro-5-methylphenol is a fluorinated aromatic compound.

Application

2-Fluoro-5-methylphenol may be employed as additive in various alkyne metatheses.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

152.6 °F - closed cup

flash_point_c

67 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkyne metathesis.
Furstner A and Davies PW.
Chemical Communications (Cambridge, England), 18, 2307-2320 (2005)
19F-encoded combinatorial libraries: discovery of selective metal binding and catalytic peptoids.
Pirrung MC, et al.
Journal of Combinatorial Chemistry, 4(4), 329-344 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service