Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

469602

Sigma-Aldrich

4-Benzyloxybutyric acid

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$115.90
25 ML
$382.85

$115.90

List Price$122.00
Web-Only Promotion

Available to ship onApril 15, 2025Details


Request a Bulk Order

Select a Size

Change View
5 ML
$115.90
25 ML
$382.85

About This Item

Linear Formula:
C6H5CH2O(CH2)3CO2H
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$115.90

List Price$122.00
Web-Only Promotion

Available to ship onApril 15, 2025Details


Request a Bulk Order

Quality Level

assay

95%

refractive index

n20/D 1.512 (lit.)

bp

135 °C/0.3 mmHg (lit.)

density

1.097 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ether
phenyl

SMILES string

OC(=O)CCCOCc1ccccc1

InChI

1S/C11H14O3/c12-11(13)7-4-8-14-9-10-5-2-1-3-6-10/h1-3,5-6H,4,7-9H2,(H,12,13)

InChI key

CXEFZVVLTJQWBF-UHFFFAOYSA-N

Related Categories

General description

4-Benzyloxybutyric acid can be synthesized from γ-butyrolactone.[1] It participates in the asymmetric total synthesis of erythromycin.[2]

Application

4-Benzyloxybutyric acid may be used in the synthesis of benzyloxybutyryl (BOB) esters of alcohols by standard acylation techniques or by the Jacobsen asymmetric nucleophilic ring opening of epoxides.[3]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric total synthesis of erythromcin. 1. Synthesis of an erythronolide A secoacid derivative via asymmetric induction.
Woodward RB, et al.
Journal of the American Chemical Society, 103(11), 3210-3213 (1981)
What about BOB? A synthetically useful protecting group.
Clark MA and Ganem B.
Tetrahedron Letters, 41(49), 9523-9526 (2000)
Synthesis of tylonolide, the aglycone of tylosin.
Masamune S, et al.
Journal of the American Chemical Society, 104(20), 5523-5526 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service