Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46877

Sigma-Aldrich

2-Fluoro-3,3-dimethyl-2,3-dihydro-1,2-benzisothiazole 1,1-dioxide

≥98.0% (F)

Synonym(s):

2,3-Dihydro-3,3-dimethyl-2-fluoro-1,2-benzisothiazole 1,1-dioxide, N-Fluoro-2,α-cumenesultam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10FNO2S
CAS Number:
Molecular Weight:
215.24
Beilstein/REAXYS Number:
4746973
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (F)

form

solid

mp

115-117 °C

solubility

THF: soluble(lit.)
acetonitrile: soluble(lit.)
diethyl ether: soluble(lit.)
organic solvents: soluble(lit.)

SMILES string

CC1(C)N(F)S(=O)(=O)c2ccccc12

InChI

1S/C9H10FNO2S/c1-9(2)7-5-3-4-6-8(7)14(12,13)11(9)10/h3-6H,1-2H3

InChI key

VMDJWSACUFRJRE-UHFFFAOYSA-N

Other Notes

Convenient reagent for electrophilic fluorination[1][2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

E. Differding et al.
Tetrahedron Letters, 32, 1779-1779 (1991)
E. Differding et al.
Tetrahedron Letters, 32, 3819-3819 (1991)
Simon E Ward et al.
Journal of medicinal chemistry, 54(1), 78-94 (2010-12-07)
A novel series of AMPAR positive modulators is described that were identified by high throughput screening. The molecules of the series have been optimized from a high quality starting point hit to afford excellent developability, tolerability, and efficacy profiles, leading

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service