Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

468657

Sigma-Aldrich

2-Chloroethanol

99%

Synonym(s):

Ethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2OH
CAS Number:
Molecular Weight:
80.51
Beilstein/REAXYS Number:
878139
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

2.78 (vs air)

vapor pressure

5 mmHg ( 20 °C)

assay

99%

autoignition temp.

797 °F

expl. lim.

16 %

refractive index

n20/D 1.441 (lit.)

bp

129 °C (lit.)

mp

−89 °C (lit.)

density

1.201 g/mL at 25 °C (lit.)

SMILES string

OCCCl

InChI

1S/C2H5ClO/c3-1-2-4/h4H,1-2H2

InChI key

SZIFAVKTNFCBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in double-wall stainless steel tanks

Replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service