462225
Tri-O-acetyl-D-galactal
97%
Synonym(s):
1,2-Dideoxy-3,4,6-tri-O-acetyl-D-lyxo-1-hexenopyranose
About This Item
Recommended Products
assay
97%
optical activity
[α]22/D −18°, c = 1 in chloroform
mp
34-38 °C (lit.)
SMILES string
CC(=O)OC[C@H]1OC=C[C@@H](OC(C)=O)[C@H]1OC(C)=O
InChI
1S/C12H16O7/c1-7(13)17-6-11-12(19-9(3)15)10(4-5-16-11)18-8(2)14/h4-5,10-12H,6H2,1-3H3/t10-,11-,12-/m1/s1
InChI key
LLPWGHLVUPBSLP-IJLUTSLNSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service