Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

458368

Sigma-Aldrich

Poly(1,6-hexamethylene adipate)

average Mn ~3,800

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-O(CH2)6O2C(CH2)4CO-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002

mol wt

average Mn ~3,800

viscosity

4,200(60 °C)(lit.)

bp

>400 °C (lit.)

hydroxyl functionality

2.0

hydroxyl value

30 mg KOH/g

transition temp

Tm 55-65 °C

density

1.13 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCO.OC(=O)CCCCC(O)=O

Packaging

Packaged in poly bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service