Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

454036

Sigma-Aldrich

Aurintricarboxylic acid ammonium salt

Synonym(s):

ATA, Aluminon, Ammonium aurintricarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H14O9 · 3NH3
CAS Number:
Molecular Weight:
473.43
Colour Index Number:
43810
Beilstein/REAXYS Number:
3900820
EC Number:
MDL number:
UNSPSC Code:
12171500

mp

220-225 °C (dec.) (lit.)

λmax

450 nm
522 nm (2nd)

SMILES string

N.N.N.OC(=O)c1cc(ccc1O)\C(c2ccc(O)c(c2)C(O)=O)=C3/C=CC(=O)C(=C3)C(O)=O

InChI

1S/C22H14O9.3H3N/c23-16-4-1-10(7-13(16)20(26)27)19(11-2-5-17(24)14(8-11)21(28)29)12-3-6-18(25)15(9-12)22(30)31;;;/h1-9,23-24H,(H,26,27)(H,28,29)(H,30,31);3*1H3

InChI key

AIPNSHNRCQOTRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Aurintricarboxylic acid readily polymerizes in aqueous solution, forming a stable free radical that inhibits protein-nucleic acid interactions. It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.

Other Notes

May contain a substantial amount of polymeric material.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service