Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446815

Sigma-Aldrich

2,2,3,3-Tetrafluoro-1,4-butanediol

98%

Synonym(s):

2,2,3,3-Tetrafluorobutanediol, NSC 95113, Tetrafluoro-1,4-butanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CF2CF2CH2OH
CAS Number:
Molecular Weight:
162.08
MDL number:
UNSPSC Code:
12352100

assay

98%

bp

110-112 °C/13 mmHg (lit.)

mp

77-82 °C (lit.)
85-87 °C

SMILES string

OCC(F)(F)C(F)(F)CO

InChI

1S/C4H6F4O2/c5-3(6,1-9)4(7,8)2-10/h9-10H,1-2H2

InChI key

CDZXJJOGDCLNKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the preparation of polyfluorosiloxanes as precursors to per- and polyfluoroethers.
Reactant for:
Enzymic synthesis of silicone fluorinated aliphatic polyester amides

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic Chemistry, 33, 415-415 (1994)
Inorganic Chemistry, 33, 5463-5463 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service