Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444057

Sigma-Aldrich

Ethyltriacetoxysilane

95%

Synonym(s):

Triacetoxy(ethyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3SiC2H5
CAS Number:
Molecular Weight:
234.28
Beilstein/REAXYS Number:
1795013
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

refractive index

n20/D 1.411 (lit.)

bp

98 °C/4 mmHg (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CC[Si](OC(C)=O)(OC(C)=O)OC(C)=O

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service