Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442097

Sigma-Aldrich

Platinum(II) cyanide

99.99% trace metals basis

Synonym(s):

Platinum dicyanide, Platinum(II) cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(CN)2
CAS Number:
Molecular Weight:
247.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99.99% trace metals basis
form:
solid

assay

99.99% trace metals basis

form

solid

SMILES string

N#C[Pt]C#N

InChI

1S/2CN.Pt/c2*1-2;

InChI key

DEUYCYIBUSRJFZ-UHFFFAOYSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

supp_hazards

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service