Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441201

Sigma-Aldrich

3,4-Difluoroiodobenzene

98%

Synonym(s):

1,2-Difluoro-4-iodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3I
CAS Number:
Molecular Weight:
239.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

contains

copper as stabilizer

refractive index

n20/D 1.558 (lit.)

bp

177-178 °C (lit.)

density

1.99 g/mL at 25 °C (lit.)

Storage temp.

2-8°C

SMILES string

Fc1ccc(I)cc1F

InChI

1S/C6H3F2I/c7-5-2-1-4(9)3-6(5)8/h1-3H

Inchi Key

KSASJELKLBIMSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Difluoroiodobenzene is an aryl fluorinated building block.

Application

3,4-Difluoroiodobenzene may be employed for the functionalization of Si substrate, via Heck arylation and alkyl Suzuki coupling reaction.[1]

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

163.4 °F - closed cup

flash_point_c

73 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Medicinal Chemistry, 39, 2780-2780 (1996)
Gillian Collins et al.
Langmuir : the ACS journal of surfaces and colloids, 29(38), 11950-11958 (2013-08-24)
Palladium-catalyzed Suzuki, Heck, and Sonogashira coupling reactions were studied as reaction protocols for organic modification of Si surfaces. These synthetically useful protocols allow for surface modification of alkene, alkyne, and halide terminated surfaces. Surface oxidation and metal contamination were assessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service