Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439959

Sigma-Aldrich

4-Isopropylphenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4NCO
CAS Number:
Molecular Weight:
161.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

98%

refractive index

n20/D 1.52 (lit.)

bp

85 °C/4 mmHg (lit.)

density

1.01 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccc(cc1)N=C=O

InChI

1S/C10H11NO/c1-8(2)9-3-5-10(6-4-9)11-7-12/h3-6,8H,1-2H3

InChI key

PJVRNNRZWASOIT-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service