Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

438391

Sigma-Aldrich

Pyridinium teflate

97%

Synonym(s):

Pyridinium pentafluorooxotellurate(VI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6F5NOTe
CAS Number:
Molecular Weight:
318.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

mp

211.5 °C (dec.) (lit.)

SMILES string

c1cc[nH+]cc1.F[Te-](F)(F)(F)(F)=O

InChI

1S/C5H5N.F5HOTe/c1-2-4-6-5-3-1;1-7(2,3,4,5)6/h1-5H;6H

InChI key

MGPSFNJAJSYBQN-UHFFFAOYSA-N

General description

Pyridinium teflate is a pyridinium salt of OTeF5.[1]

Application

Pyridinium teflate is a stable starting material for vacuum line production of teflic acid, a very weakly coordinating ligand.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electronic and molecular structure of pentafluorooxotellurate (1-).
Miller PK, et al.
Inorganic Chemistry, 27(13), 2255-2261 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service