Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436313

Sigma-Aldrich

Octadecyl vinyl ether

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17OCH=CH2
CAS Number:
Molecular Weight:
296.53
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

assay

85%

impurities

<15% hexadecyl vinyl ether

bp

183-192 °C/7 mmHg (lit.)

mp

24-28 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOC=C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service