Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434132

Sigma-Aldrich

tert-Butyl methyl-d3 ether

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCD3
CAS Number:
Molecular Weight:
91.17
Beilstein/REAXYS Number:
2036450
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.368 (lit.)

bp

55-56 °C (lit.)

density

0.765 g/mL at 25 °C (lit.)

application(s)

microbiology

mass shift

M+3

SMILES string

[2H]C([2H])([2H])OC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i4D3

InChI key

BZLVMXJERCGZMT-GKOSEXJESA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl-d3 ether is a deuterated NMR solvent useful in NMR-based research and analyses.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

14.0 °F - closed cup

flash_point_c

-10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service