Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

434078

Sigma-Aldrich

Erbium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate)

≥99.9% trace metals basis

Synonym(s):

Er(TMHD)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Er(OCC(CH3)3CHCOC(CH3)3)3
CAS Number:
Molecular Weight:
717.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

≥99.9% trace metals basis

form

powder or crystals

reaction suitability

core: erbium
reagent type: catalyst

SMILES string

CC(C)(C)C(=O)\C=C(/O[Er](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/3C11H20O2.Er/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b3*8-7-;

InChI key

PRYGICXWMYVFKG-LWTKGLMZSA-K

General description

Atomic number of base material: 68 Erbium

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

High Purity Metalorganic Precursors for CPV Device Fabrication

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service