Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43156

Sigma-Aldrich

(2S,4S)-(−)-4-Diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine

≥98.0% (CHN)

Synonym(s):

PPM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H29NP2
CAS Number:
Molecular Weight:
453.49
Beilstein/REAXYS Number:
500619
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

≥98.0% (CHN)

optical activity

[α]20/D −15.5±1°, c = 1% in benzene

mp

102-104 °C (lit.)
102-104 °C

SMILES string

C1N[C@@H](C[C@@H]1P(c2ccccc2)c3ccccc3)CP(c4ccccc4)c5ccccc5

InChI

1S/C29H29NP2/c1-5-13-25(14-6-1)31(26-15-7-2-8-16-26)23-24-21-29(22-30-24)32(27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-20,24,29-30H,21-23H2/t24-,29-/m0/s1

InChI key

PQHKXSOICTYGQD-OUTSHDOLSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral diphosphine for asymmetric hydrogenations; starting material for the synthesis of various N-substituted diphosphines[1][2][3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J DiNapoli et al.
PloS one, 14(1), e0210409-e0210409 (2019-01-11)
Explaining the processes underlying the emergence of monument construction is a major theme in contemporary anthropological archaeology, and recent studies have employed spatially-explicit modeling to explain these patterns. Rapa Nui (Easter Island, Chile) is famous for its elaborate ritual architecture
C. Yang et al.
Organometallics, 20, 5220-5220 (2001)
Roman Brenner et al.
Europace : European pacing, arrhythmias, and cardiac electrophysiology : journal of the working groups on cardiac pacing, arrhythmias, and cardiac cellular electrophysiology of the European Society of Cardiology, 19(7), 1220-1226 (2016-10-06)
Elderly patients with sinus node dysfunction (SND) are at increased risk of falls with possible injuries. However, the incidence of these adverse events and its reduction after permanent pacemaker (PPM) implantation are not known. Eighty-seven patients (mean [SD] age 75.4
Dmitry Sutormin et al.
Nucleic acids research, 47(3), 1373-1388 (2018-12-06)
An important antibiotic target, DNA gyrase is an essential bacterial enzyme that introduces negative supercoils into DNA and relaxes positive supercoils accumulating in front of moving DNA and RNA polymerases. By altering the superhelical density, gyrase may regulate expression of
Nasrin Joudyian et al.
BMC health services research, 21(1), 4-4 (2021-01-06)
The Astana Declaration on Primary Health Care reiterated that PHC is a cornerstone of a sustainable health system for universal health coverage (UHC) and health-related Sustainable Development Goals. It called for governments to give high priority to PHC in partnership

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service