Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

427705

Sigma-Aldrich

3-(2-Chloroethyl)-2,4(1H,3H)-quinazolinedione

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9ClN2O2
CAS Number:
Molecular Weight:
224.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

mp

195-197 °C (lit.)

functional group

chloro

SMILES string

ClCCN1C(=O)Nc2ccccc2C1=O

InChI

1S/C10H9ClN2O2/c11-5-6-13-9(14)7-3-1-2-4-8(7)12-10(13)15/h1-4H,5-6H2,(H,12,15)

InChI key

HWFSVCPXNLEACG-UHFFFAOYSA-N

General description

3-(2-Chloroethyl)-2,4(1H,3H)-quinazolinedione is a quinazolinedione derivative. It has been synthesized by reacting ethyl-2-aminobenzoate with ethyl chloroformate and ethanol amine.[1]

Application

3-(2-Chloroethyl)-2,4(1H,3H)-quinazolinedione may be used in the synthesis of ketanserin .[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reinvestigation of the Synthesis of Ketanserin (5) and its Hydrochloride Salt (5. HCl) via 3-(2-Chloroethyl)-2,4-(1H,3H)-quinazolinedione (2) or Dihydro-5H-oxazole (2,3-b) quinazolin-5-one (1).
Fakhraian H and Heydary M.
Journal of Heterocyclic Chemistry, 51(1), 151-156 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service