Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

424986

Sigma-Aldrich

2-(2-Oxo-1-imidazolidinyl)ethyl methacrylate solution

25 wt. % in methyl methacrylate

Synonym(s):

1-(2-Methacryloyloxyethyl)-2-imidazolidinone, 2-(2-Oxoimidazolidin-1-yl)ethyl methacrylate, N -(2-Methacryloxyethyl)imidazolidin-2-one, N -[2-Methacryloyloxyethyl]ethyleneurea, Methacryloxyethyl ethylene urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N2O3
CAS Number:
Molecular Weight:
198.22
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

liquid

contains

225-275 ppm phenothiazine as inhibitor
70-110 ppm monomethyl ether hydroquinone as inhibitor

concentration

25 wt. % in methyl methacrylate

refractive index

n20/D 1.437

bp

100 °C

mp

−30 °C

density

0.99 g/mL at 25 °C

SMILES string

CC(=C)C(=O)OCCN1CCNC1=O

InChI

1S/C9H14N2O3/c1-7(2)8(12)14-6-5-11-4-3-10-9(11)13/h1,3-6H2,2H3,(H,10,13)

InChI key

PFPUZMSQZJFLBK-UHFFFAOYSA-N

Application

Adhesion promoter

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service