Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

421979

Sigma-Aldrich

Bis(hexamethylene)triamine

technical grade, 60%

Synonym(s):

6,6′-Iminodihexylamine, Bis(6-aminohexyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)6NH(CH2)6NH2
CAS Number:
Molecular Weight:
215.38
Beilstein/REAXYS Number:
2323517
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical grade

vapor pressure

<0.01 mmHg ( 25 °C)

concentration

60%

refractive index

n20/D 1.49 (lit.)

bp

163-165 °C/4 mmHg (lit.)

mp

33-36 °C (lit.)

density

0.85 g/mL at 20 °C (lit.)
0.926 g/mL at 25 °C

SMILES string

NCCCCCCNCCCCCCN

InChI

1S/C12H29N3/c13-9-5-1-3-7-11-15-12-8-4-2-6-10-14/h15H,1-14H2

InChI key

MRNZSTMRDWRNNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service