Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417394

Sigma-Aldrich

1-(2-Hydroxyphenyl)-3-phenyl-1,3-propanedione

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COCH2COC6H5
CAS Number:
Molecular Weight:
240.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

120-122 °C (lit.)

functional group

ketone
phenyl

SMILES string

Oc1ccccc1C(=O)CC(=O)c2ccccc2

InChI

1S/C15H12O3/c16-13-9-5-4-8-12(13)15(18)10-14(17)11-6-2-1-3-7-11/h1-9,16H,10H2

InChI key

OABFIJGAEVKMJP-UHFFFAOYSA-N

General description

1-(2-Hydroxyphenyl)-3-phenyl-1,3-propanedione is a chalcone. It has been isolated from the stem bark of Millettia ovalifolia and the root bark of Pongamia pinnata. It can be prepared by the benzoylation of ortho-hydroxyacetophenone in pyridine. It has been reported to crystallize in the triclinic space group. It exists in two enolic tautomeric forms in which the hydroxyl and phenolic protons are linked to the ketonic oxygen atom via hydrogen bonds.[1] It forms (1:1) inclusion compounds by reacting with formaldehyde in the presence of various 1-arylpiperazines.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abnormal base catalysed reaction of formaldehyde and benzaldehydes with 1-(2-hydroxyphenyl)-3-phenyl-1, 3-propanedione.
Joglekar SJ and Samant SD.
Tetrahedron Letters, 29(2), 241-244 (1988)
Crystal structures of 1-(2-hydroxyphenyl)-3-phenyl-1, 3-propanedione and 1-(1, 3-benzodioxol-5-yl)-3-(2, 4-dimethoxyphenyl)-1, 3-propanedione.
Ng SW, et al.
Journal of Chemical Crystallography, 24(5), 331-335 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service