Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41470

Sigma-Aldrich

cis-2,6-Dimethylpiperidine

≥97.0% (GC)

Synonym(s):

2,6-Lupetidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N
CAS Number:
Molecular Weight:
113.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.4394 (lit.)
n20/D 1.440

bp

127-128 °C/768 mmHg (lit.)

density

0.84 g/mL at 25 °C (lit.)

SMILES string

C[C@H]1CCC[C@@H](C)N1

InChI

1S/C7H15N/c1-6-4-3-5-7(2)8-6/h6-8H,3-5H2,1-2H3/t6-,7+

Inchi Key

SDGKUVSVPIIUCF-KNVOCYPGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

cis-2,6-Dimethylpiperidine [cis-DMP(H)] may be used for the preparation of four new zincates and one new magnesiate, where [TMEDA = N,N,N´,N´-tetramethylethylenediamine; cis-DMP = cis-2,6-dimethylpiperidide; DIBA(H) = diisobutylamine]:[1]
  • (TMEDA)·LiZn(cis-DMP)(t Bu)2
  • (TMEDA)·NaZn(cis-DMP)(t Bu)2
  • (TMEDA)·NaZn(cis-DMP)2(t Bu)
  • (TMEDA)·NaZn(DIBA)2(t Bu)
  • (TMEDA)·NaMg(cis-DMP)3
Reagent for elimination reactions

Reactant for corrosion inhibitors for iron in HCl

Catalyst for:
Preparation of aliphatic amines
Ketene forming eliminations
Stereoselective synthesis of triflates

Pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proton magnetic resonance studies of cyclic compounds-VI: Cis-and trans-2, 6-dimethylpiperidine and cis (2, 4), cis (4, 6), cis (2, 6)-2, 4, 6-trimethylpiperidine.
Booth HJ< et al.
Tetrahedron, 24(1), 279-287 (1968)
Ross Campbell et al.
Dalton transactions (Cambridge, England : 2003), (2)(2), 511-519 (2009-12-22)
Four novel heterobimetallic ate complexes containing cis-2,6-dimethylpiperidide (cis-DMP) have been prepared and characterised. Two contain one cis-DMP ligand, namely the bisalkyl-amido lithium, and sodium zincates [(TMEDA) x MZn(cis-DMP)(tBu)2] (M = Li for 1, Na for 2). Both 1 and 2
Anton Vidal-Ferran et al.
The Journal of organic chemistry, 63(18), 6309-6318 (2001-10-24)
Enantiomerically pure (2S,3S)-2,3-epoxy-3-phenylpropanol (3) has been anchored to Merrifield resins with different degrees of cross-linking and functionalization. The resulting epoxy-functionalized resins 4 have been submitted to completely regioselective (C-3 attack) and stereospecific ring-opening with secondary amines [piperidine (a), N-methylpiperazine (b)
Dawid Jędrzkiewicz et al.
Polymers, 12(9) (2020-09-04)
An efficient synthesis strategy of a well-defined polylactide-dye conjugate in a controlled fashion is presented. The introduction of coloring species as end groups of polylactide (PLA) has been performed by using new homoleptic aminophenolate magnesium or zinc coordination compounds. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service