409804
4-Chloro-7-sulfobenzofurazan ammonium salt
97%
Synonym(s):
4-Chloro-7-sulfobenz-2-oxa-1,3-diazole ammonium salt, 7-Chloro-2,1,3-benzoxadiazole-4-sulfonic acid ammonium salt, 7-Chloro-4-benzofurazansulfonic acid ammonium salt, SBF-Chloride
About This Item
assay
97%
mp
>300 °C (lit.)
SMILES string
N.OS(=O)(=O)c1ccc(Cl)c2nonc12
InChI
1S/C6H3ClN2O4S.H3N/c7-3-1-2-4(14(10,11)12)6-5(3)8-13-9-6;/h1-2H,(H,10,11,12);1H3
InChI key
QFHXPBLOMHGYOC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service