Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40825

Sigma-Aldrich

3-Hydroxy-1,2,3-benzotriazin-4(3H)-one solution

~25% in DMF

Synonym(s):

3,4-Dihydro-3-hydroxy-4-oxo-1,2,3-benzotriazine, DHBT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O2
CAS Number:
Molecular Weight:
163.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

liquid

concentration

~25% in DMF

impurities

≤0.1% water

refractive index

n20/D 1.477

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

ON1N=Nc2ccccc2C1=O

InChI

1S/C7H5N3O2/c11-7-5-3-1-2-4-6(5)8-9-10(7)12/h1-4,12H

InChI key

HJBLUNHMOKFZQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Solution may crystallize when stored over extended periods of time. Gently warm to ambient temperature and stir to bring product back into solution

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

136.4 °F - closed cup

flash_point_c

58 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service