Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

408204

Sigma-Aldrich

Chlorodicyclohexylborane solution

1.0 M in methylene chloride

Synonym(s):

Dicyclohexylboron chloride, Dicyclohexylboryl chloride, Dicyclohexylchloroborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2BCl
CAS Number:
Molecular Weight:
212.57
Beilstein/REAXYS Number:
3233942
MDL number:
UNSPSC Code:
12352000

concentration

1.0 M in methylene chloride

SMILES string

ClB(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22BCl/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

PBBOKJIYEZCTEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Aldol addition reactions
  • Mukaiyama aldol addition
  • Microwave assisted ring closing metathesis reactions
  • C-alkylation of aromatic aldimines
  • Reversible ketone-ketone aldol reactions
  • Syn- and anti-stereoselective aldol reaction hexenone with aldehydes

Packaging

Packaged under nitrogen in Sure/Seal bottles

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service