Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

407747

Sigma-Aldrich

1,2-Epoxypentane

98%

Synonym(s):

Propyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

98%

refractive index

n20/D 1.396 (lit.)

bp

89-90 °C/750 mmHg (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CCCC1CO1

InChI

1S/C5H10O/c1-2-3-5-4-6-5/h5H,2-4H2,1H3

InChI key

SYURNNNQIFDVCA-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

23.0 °F - closed cup

flash_point_c

-5 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service