Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

406759

Sigma-Aldrich

Poly[(phenyl glycidyl ether)-co-formaldehyde]

average Mn ~605

Synonym(s):

Phenol-formaldehyde polymer glycidyl ether, Phenol-formaldehyde polymer, oxiranylmethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
Pricing and availability is not currently available.

specific activity

3.8 epoxide groups per molecule

mol wt

average Mn ~605

transition temp

softening point 48-58 °C

density

1.227 g/mL at 25 °C (lit.)

SMILES string

[H]C([H])=O.C1OC1COc2ccccc2

Looking for similar products? Visit Product Comparison Guide

Application

Adhesives, structural and electrical laminates and coatings and castings for high temperature service.

Features and Benefits

Good chemical resistance at high temperatures. Substitutes for Bisphenol A and phenolic epoxy resins.

Physical form

Oligomeric epoxide.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service