Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40440

Sigma-Aldrich

N,N-Dimethylheptylamine

≥97.0%

Synonym(s):

1-(Dimethylamino)heptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6N(CH3)2
CAS Number:
Molecular Weight:
143.27
Beilstein/REAXYS Number:
1697547
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97.0%

refractive index

n20/D 1.421

bp

60 °C/10 mmHg (lit.)

density

0.757 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCN(C)C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service