Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

400882

Sigma-Aldrich

Lithium dichromate hydrate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cr2Li2O7 · xH2O
CAS Number:
Molecular Weight:
229.87 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

technical grade

composition

Cr(VI), 41%

reaction suitability

reagent type: oxidant

SMILES string

[Li+].[Li+].O.[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

signalword

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Muta. 1B - Ox. Sol. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service