Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394823

Sigma-Aldrich

2,2-Bis(trifluoromethyl)-1,3-dioxolane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4F6O2
CAS Number:
Molecular Weight:
210.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.312 (lit.)

bp

106-107 °C (lit.)

density

1.53 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C1(OCCO1)C(F)(F)F

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service