About This Item
Recommended Products
grade
technical grade
assay
90%
refractive index
n20/D 1.512 (lit.)
bp
142-144 °C (lit.)
density
1.905 g/mL at 25 °C (lit.)
SMILES string
CCC(Br)C(Br)=O
InChI
1S/C4H6Br2O/c1-2-3(5)4(6)7/h3H,2H2,1H3
InChI key
HHKDBXNYWNUHPL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
target_organs
Respiratory system
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service