Skip to Content
MilliporeSigma
All Photos(1)

Documents

388807

Sigma-Aldrich

3-(N,N-Dimethylpalmitylammonio)propanesulfonate

97%

Synonym(s):

3-(N,N-Dimethylhexadecylammonio)propanesulfonate, 3-(Palmityldimethylammonio)propanesulfonate, N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, Palmityl sulfobetaine, SB3-16, Zwittergent® 3-16

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
391.65
Beilstein/REAXYS Number:
4149854
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

SMILES string

CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C21H45NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22(2,3)20-18-21-26(23,24)25/h4-21H2,1-3H3

InChI key

TUBRCQBRKJXJEA-UHFFFAOYSA-N

Legal Information

Zwittergent is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan Cao et al.
Journal of chromatography. A, 1358, 277-284 (2014-07-23)
A rapid zwitterionic microemulsion electrokinetic chromatography (ZI-MEEKC) approach coupled with light-emitting-diode-induced fluorescence (LED-IF, 480nm) detection was proposed for the analysis of flavonoids. In the optimization process, we systematically investigated the separation conditions, including the surfactants, cosurfactants, pH, buffers and fluorescence
Francisco J Prado-Prado et al.
Bioorganic & medicinal chemistry, 18(6), 2225-2231 (2010-02-27)
There are many of pathogen parasite species with different susceptibility profile to antiparasitic drugs. Unfortunately, almost QSAR models predict the biological activity of drugs against only one parasite species. Consequently, predicting the probability with which a drug is active against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service