Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

386332

Sigma-Aldrich

7,9-Dimethylbenz[c]acridine

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15N
CAS Number:
Molecular Weight:
257.33
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

bp

300 °C/10 mmHg (lit.)

mp

157-161 °C (lit.)

SMILES string

Cc1ccc2nc3c(ccc4ccccc34)c(C)c2c1

InChI

1S/C19H15N/c1-12-7-10-18-17(11-12)13(2)15-9-8-14-5-3-4-6-16(14)19(15)20-18/h3-11H,1-2H3

InChI key

HEFJMRLDXHSXEP-UHFFFAOYSA-N

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Ye et al.
Carcinogenesis, 16(4), 787-793 (1995-04-01)
The major and minor metabolites of the potent polycyclic aza-aromatic carcinogens 7,9-dimethylbenz[c]acridine and 7,10-dimethylbenz[c]acridine, and the stereochemistry of the dihydrodiol metabolites have been previously described. The metabolite distributions produced in incubations of the aza-aromatic compounds with liver microsomes from phenobarbital-
Effects of several dimethylbenzacridines on secondary hamster embryo cells: neoplastic transformation.
D Papadopoulo et al.
European journal of cancer, 17(2), 179-186 (1981-02-01)
Y Ye et al.
Chemical research in toxicology, 8(2), 188-202 (1995-03-01)
The hepatic microsomal metabolites of the highly carcinogenic dimethylbenzacridines, 7,9-dimethylbenz[c]acridine (7,9-DMBAC), and 7,10-dimethylbenz[c]acridine (7,10-DMBAC) were obtained with preparations from 3-methylcholanthrene-pretreated rats. Metabolites were separated by reversed-phase HPLC and characterized using UV spectral data and chemical ionization-mass spectrometry after trimethylsilylation and
Y Ye et al.
Chemical research in toxicology, 8(2), 203-208 (1995-03-01)
The monofunctionalized dihydrodiol metabolites of 7,9-dimethylbenz[c]acridine and 7,10-dimethylbenz[c]acridine formed in incubations with rat liver microsomes from untreated and phenobarbital and 3-methylcholanthrene-pretreated rats were isolated by reversed-phase high performance liquid chromatography. The relative amounts of each enantiomer were determined by HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service