Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38223

Sigma-Aldrich

Diisooctylphosphinic acid

technical, ~90% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H35O2P
CAS Number:
Molecular Weight:
290.42
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

~90% (T)

form

liquid

refractive index

n20/D 1.460

density

0.916 g/mL at 20 °C (lit.)

SMILES string

CC(C)CCCCCP(O)(=O)CCCCCC(C)C

InChI

1S/C16H35O2P/c1-15(2)11-7-5-9-13-19(17,18)14-10-6-8-12-16(3)4/h15-16H,5-14H2,1-4H3,(H,17,18)

InChI key

GBNVBFGHGMAMDH-UHFFFAOYSA-N

General description

Diisooctylphosphinic acid is an organophosphorous acid.

Application

Diisooctylphosphinic acid (DiOPA) may be used in the following studies:
  • As the surface passivating ligand in the synthesis of magic-sized CdSe and CdTe nanocrystals.[1]
  • As extractant in the extraction of Indium(III) from sulfate solutions.[2]
  • As ligand to investigate the solubility of four copper ligand systems in supercritical CO2.[3]
  • As acid ligand for the Supercritical fluid extraction (SFE) of divalent metals (Zn2+, Cu2+, Pb2+, Cd2+ and Cr3+) from sand and fly ash using CO2.[4]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of magic-sized CdSe and CdTe nanocrystals with diisooctylphosphinic acid.
Dukes III AD, et al.
Chemistry of Materials, 22(23), 6402-6408 (2010)
Extraction of heavy metals from fly ash and sand with ligands and supercritical carbon dioxide.
Kersch C, et al.
Industrial & Engineering Chemistry Research, 39(12), 4670-4672 (2000)
De la Rosa JR, et al.
Materials and Manufacturing Processes, 26(2), 304-310 (2011)
Extraction of indium (III) from sulfate solutions with organophosphorus acids.
Travkin VF, et al.
Russian Journal of Applied Chemistry, 77(10), 1613-1617 (2004)
Taeyjuana Y Lyons et al.
Langmuir : the ACS journal of surfaces and colloids, 33(12), 3018-3027 (2017-03-01)
The increasing commercialization of consumer electronic products that make use of II-VI semiconductor quantum dots (QDs) has raised significant concerns about their impact on natural systems and human health once they are released into the environment at the end of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service