Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

380059

Sigma-Aldrich

4-Chloro-3-nitrobenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CN
CAS Number:
Molecular Weight:
182.56
Beilstein/REAXYS Number:
1639111
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

98-100 °C (lit.)

functional group

chloro
nitrile
nitro

SMILES string

[O-][N+](=O)c1cc(ccc1Cl)C#N

InChI

1S/C7H3ClN2O2/c8-6-2-1-5(4-9)3-7(6)10(11)12/h1-3H

InChI key

XBLPHYSLHRGMNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

FT-IR and μ-Raman spectra of 4-chloro-3-nitrobenzonitrile have been recorded in the range of 400-4000cm-1 and 100-4000cm-1, respectively.[1] It has been prepred from the reaction of 4-chloro-3-nitrobenzamide and phosphorus oxychloride. Crystal structure of 4-chloro-3-nitrobenzonitrile has been reported. Molecules of 4-chloro-3-nitrobenzonitrile are linked by weak intermolecular C-H…O and C-H…N hydrogen bonds.[2]

Application

4-Chloro-3-nitrobenzonitrile may be employed for the radiosynthesis of analogs of serotonin transporter (SERT) ligands for use in positron emission tomography (PET) studies[3] and for the preparation of N-(4-cyano-2-nitrophenyl)glycine.[4]
4-Chloro-3-nitrobenzonitrile may be used in the synthesis of 3-nitro-4-thiocyanobenzonitryl.[5]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Galina R Demina et al.
PloS one, 4(12), e8174-e8174 (2009-12-18)
Resuscitation promoting factors (RPF) are secreted proteins involved in reactivation of dormant actinobacteria, including Mycobacterium tuberculosis. They have been considered as prospective targets for the development of new anti-tuberculosis drugs preventing reactivation of dormant tubercle bacilli, generally associated with latent
Bo-Nian Liu et al.
Acta crystallographica. Section E, Structure reports online, 65(Pt 1), o92-o92 (2008-01-01)
In the title compound, C(7)H(3)ClN(2)O(2), the Cl, C and N atoms are coplanar with the aromatic ring. In the crystal structure, weak inter-molecular C-H⋯O and C-H⋯N hydrogen bonds link the mol-ecules. The π-π contact between the benzene rings, [centroid-centroid distances
S X Cai et al.
Journal of medicinal chemistry, 40(5), 730-738 (1997-02-28)
We report on a series of alkyl- and alkoxy-substituted 1,4-dihydroquinoxaline-2,3-diones (QXs), prepared as a continuation of our structure-activity relationship (SAR) study of QXs as antagonists for the glycine site of the N-methyl-D-aspartate (NMDA) receptor. The in vitro potency of these
A A Wilson et al.
Journal of medicinal chemistry, 43(16), 3103-3110 (2000-08-24)
A series of four 2-(phenylthio)araalkylamines have been radiolabeled with (11)C and evaluated as potential radiotracers for imaging the serotonin transporter (SERT) by positron emission tomography (PET). All four candidates display high affinity for SERT and low affinity for the dopamine
Yusuf Sert et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 107, 248-255 (2013-02-26)
In the present study, the experimental and theoretical harmonic and anharmonic vibrational frequencies of 4-chloro-3-nitrobenzonitrile were investigated. The experimental FT-IR (400-4000 cm(-1)) and μ-Raman spectra (100-4000 cm(-1)) of the molecule in the solid phase were recorded. Theoretical vibrational frequencies and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service