Skip to Content
MilliporeSigma
All Photos(1)

Documents

378224

Sigma-Aldrich

3,5-Dichloro-1-fluoropyridinium triflate

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2F4NO3S
CAS Number:
Molecular Weight:
316.06
Beilstein/REAXYS Number:
4224296
MDL number:
UNSPSC Code:
12352103

grade

technical grade

assay

85%

mp

107-112 °C (lit.)

SMILES string

[O-]S(=O)(=O)C(F)(F)F.F[n+]1cc(Cl)cc(Cl)c1

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service