Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

377309

Sigma-Aldrich

Triisopropylphosphine

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]3P
CAS Number:
Molecular Weight:
160.24
Beilstein/REAXYS Number:
906732
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

technical grade

form

liquid

reaction suitability

reaction type: Cross Couplings
reagent type: ligand

refractive index

n20/D 1.466 (lit.)

bp

81 °C/22 mmHg (lit.)

density

0.839 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CC(C)P(C(C)C)C(C)C

InChI

1S/C9H21P/c1-7(2)10(8(3)4)9(5)6/h7-9H,1-6H3

InChI key

IGNTWNVBGLNYDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Pyr. Liq. 1 - Skin Corr. 1B

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

221.0 °F - closed cup

flash_point_c

105 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service