Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

376817

Sigma-Aldrich

1-(2,3,5,6-Tetrafluorophenyl)imidazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4F4N2
CAS Number:
Molecular Weight:
216.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

mp

92-94 °C (lit.)

SMILES string

Fc1cc(F)c(F)c(c1F)-n2ccnc2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Paula Pereira da Silva et al.
The Biochemical journal, 417(3), 717-726 (2008-10-24)
3-BrPA (3-bromopyruvate) is an alkylating agent with anti-tumoral activity on hepatocellular carcinoma. This compound inhibits cellular ATP production owing to its action on glycolysis and oxidative phosphorylation; however, the specific metabolic steps and mechanisms of 3-BrPA action in human hepatocellular
Jihye Yun et al.
Science (New York, N.Y.), 325(5947), 1555-1559 (2009-08-08)
Tumor progression is driven by genetic mutations, but little is known about the environmental conditions that select for these mutations. Studying the transcriptomes of paired colorectal cancer cell lines that differed only in the mutational status of their KRAS or
Axel-R Hanauske et al.
Investigational new drugs, 27(4), 356-365 (2008-10-29)
The objectives of this phase I study were to determine the maximum tolerated dose (MTD), recommended phase II dose (RD), antitumor activity, safety, and pharmacokinetics of pemetrexed-paclitaxel combination. Patients (N = 95) with advanced solid tumors were assigned to three
Ji Su Kim et al.
Journal of bioenergetics and biomembranes, 40(6), 607-618 (2008-12-11)
Hexokinase type II (HK II) is the key enzyme for maintaining increased glycolysis in cancer cells where it is overexpressed. 3-bromopyruvate (3-BrPA), an inhibitor of HK II, induces cell death in cancer cells. To elucidate the molecular mechanism of 3-BrPA-induced
Esther Hulleman et al.
Blood, 113(9), 2014-2021 (2008-11-04)
Treatment failure in pediatric acute lymphoblastic leukemia (ALL) is related to cellular resistance to glucocorticoids (eg, prednisolone). Recently, we demonstrated that genes associated with glucose metabolism are differentially expressed between prednisolone-sensitive and prednisolone-resistant precursor B-lineage leukemic patients. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service