376795
Ethyl (R)-(+)-4,4,4-trifluoro-3-hydroxybutyrate
99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
assay
99%
optical activity
[α]24/D +14.0°, c = 1 in chloroform
refractive index
n20/D 1.375 (lit.)
bp
88-90 °C/21 mmHg (lit.)
density
1.259 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)C[C@@H](O)C(F)(F)F
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
186.8 °F - closed cup
flash_point_c
86 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service